3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulfate (CHAPS) zwitterionic detergent

Supplier: G-Biosciences

Synonyms: 3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulfonate, 3-[(3-Cholamidopropyl)dimethylammonio]-1-propanesulphonate, CHAPS

DG096
71003-308EA 3169.79 USD
71003-308
3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulfate (CHAPS) zwitterionic detergent
3-[(3-Cholamidopropyl)dimethylammonio]-1-propane sulphate (CHAPS)
Formula: C₃₂H₅₈N₂O₇S
MW: 614.89 g/mol
Melting Pt: 116 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00081083
CAS Number: 75621-03-3
Merck Index: 13,02052

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR