Ampicillin sodium salt

Supplier: G-Biosciences
Danger

A051-B
71003-302EA 42.88 USD
71003-302
Ampicillin sodium salt
Ampicillin sodium salt
Formula: C₁₆H₁₈N₃NaO₄S
Storage Temperature: Refrigerator
MDL Number: MFCD00064313
CAS Number: 69-52-3
EINECS: 200-708-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR