CDI Amine Reactive Resin, G-Biosciences
Supplier: G-Biosciences
786-404
71003-152EA 234.64 USD
71003-152
CDI Amine Reactive Resin, G-Biosciences
Protein Linking and Labeling Systems
G-Biosciences' CDI Amine Reactive Agarose consists of 6% cross-linked agarose activated with CDI (1,1’-carbonyl diimidazole) to form reactive imidazole carbamates.
The activation of the resin occurs in solvent and to maintain its activity the resin is supplied in acetone to prevent hydrolysis. Upon reaction of the resin with primary amine containing molecules, i.e. proteins, in basic (pH 8.5-10) aqueous buffers the imidazole carbamates lose the imidazole group and form carbamate linkages.
G-Biosciences' CDI Amine Reactive Agarose is ideal for immobilizing peptides, small organic molecules and certain proteins and reactions can occur in organic solvent making it ideal for water-insoluble ligands.
Ordering information: Accessories included. Additional supplies available.