Immobilized D-Galactose for Lectin Purification, G-Biosciences
Supplier: G-Biosciences
786-391
71003-140EA 321.03 USD
71003-140
Immobilized D-Galactose for Lectin Purification, G-Biosciences
Protein Purification Protein Purification Resins
G-Biosciences' Immobilized D-Galactose is designed for the rapid purification and enrichment of glycoproteins, polysaccharides, glycolipids, lectins, galactosidases and other galactose-binding molecules.
G-Biosciences' Immobilized D-Galactose is also ideal for the purification of agglutinins, lectins, toxins, galactose-binding, N-acetylgalactosamine-binding or carbohydrate binding molecules.
G-Biosciences' Immobilized D-Galactose consists of agarose covalently coupled to D-galactose. The binding capacity of Immobilized D-Galactose is >20 mg Jacalin per mL resin.
Ordering information: Accessories available.