SDC™ (Steroid/Drug/Compound) Immobilization Kit, G-Biosciences
Supplier: G-Biosciences
786-271
71003-104EA 848.8 USD
71003-104
SDC™ (Steroid/Drug/Compound) Immobilization Kit, G-Biosciences
Protein Linking and Labeling Systems
G-Biosciences' SDC™ (Steroid/ Drug/ Compound) Immobilization kit is designed for the immobilization of steroids, drugs and chemical compounds that lack primary amines, sulfhydryls, carbonyls and other common coupling groups to a solid-phase agarose support for the use in affinity purification
G-Biosciences' SDC™ kit uses the Mannich reaction for coupling, which is described as the condensation of formaldehyde with ammonia, in the form of its salt, and another compound containing an active hydrogen. The SDC™ Immobilization kit replaces the ammonia with the primary amine on the DADPA and the active hydrogen is supplied by the steroid, drug or chemical compound to be coupled.
G-Biosciences' SDC™ kit is ideal for the generation of five 2 mL affinity columns.
The kit uses our Immobilized DADPA (diaminodipropylamine) resin that can bind steroids, drugs and chemical compounds through their active hydrogens.
Ordering information: Accessories included. Additional supplies available.