Blasticidin S hydrochloride, powder, Corning®

Supplier: Corning

30-100-RB
71002-676EA 282.44 USD
71002-676
Blasticidin S hydrochloride, powder, Corning®
Blasticidin S hydrochloride

Blasticidin S HCl is produced by Streptomyces griseochromogenes and inhibits translation in both prokaryotes and eukaryotes, thus preventing the formation of peptide bonds.

Formula: C₁₇H₂₇ClN₈O₅
Storage Temperature: Freezer
MDL Number: MFCD02091640
CAS Number: 3513-03-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR