Neomycin sulfate in solution

Supplier: IBI Scientific

Synonyms: Neomycin, Fradiomycin sulfate, (2S,3S,4R,5R,6R)-5-Amino-2-(aminomethyl)-6-(((2R,3S,4R,5S)-5-(((1R,2R,3S,5R,6S)-3,5-diamino-2-(((2R,3R,4R,5S,6R)-3-amino-6-(aminomethyl)-4,5-dihydroxytetrahydro-2H-pyran-2-yl)oxy)-6-hydroxycyclohexyl)oxy)-4-hydroxy-2-(hydroxymethyl)tetrahydrofuran-3-yl)oxy)tetrahydro-2H-pyran-3,4-diol sulphate, Fradiomycin sulphate, Neomycin sulphate salt, Neomycin sulfate salt, (2S,3S,4R,5R,6R)-5-Amino-2-(aminomethyl)-6-(((2R,3S,4R,5S)-5-(((1R,2R,3S,5R,6S)-3,5-diamino-2-(((2R,3R,4R,5S,6R)-3-amino-6-(aminomethyl)-4,5-dihydroxytetrahydro-2H-pyran-2-yl)oxy)-6-hydroxycyclohexyl)oxy)-4-hydroxy-2-(hydroxymethyl)tetrahydrofuran-3-yl)oxy)tetrahydro-2H-pyran-3,4-diol sulfate

IB02050
71002-620EA 28.9 USD
71002-620
Neomycin sulfate in solution
Neomycin sulphate

Neomycin Sulfate Solution is a clear, colorless solution used in cell culture applications


Refrigerated shipping and storage is required for this product.


It is an aminoglycoside antibiotic that inhibits translation by binding to the small subunit of prokaryotic ribosomes.

Formula: C₂₃H₄₆N₆O₁₃·3H₂SO₄
MW: 908.87 g/mol
MDL Number: MFCD00150827
CAS Number: 1405-10-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR