Polycool PG -20 Fluid, PolyScience
Supplier: POLYSCIENCE CORP-EQUIP/PARTS
060320
71002-506EA 280.16 USD
71002-506
Polycool PG -20 Fluid, PolyScience
Cleaning Agents Water Bath Protection Reagents
Propylene glycol is a safer alternative to ethylene glycol.
For use with VWR® Refrigerated Circulating Baths, VWR® Benchtop Chillers and VWR® Recirculating Chillers.
Lowers freezing point of water to allow circulation at below freezing temperatures. Mix with distilled water for broadest temperature range.