Polycool EG -25 Fluid, PolyScience
Supplier: POLYSCIENCE CORP-EQUIP/PARTS
060340
71002-504EA
135.48
USD
71002-504
Polycool EG -25 Fluid, PolyScience
Cleaning Agents
Water Bath Protection Reagents
A common circulator anti-freeze fluid.
For use with VWR® Refrigerated Circulating Baths, VWR® Benchtop Chillers and VWR® Recirculating Chillers.
Lowers the freezing point of water to allow circulation at below freezing temperatures. High toxicity. Recommend mixing with distilled water for broadest temperature range.