AMCO CLEAR® Turbidity Standard, GFS Chemicals
Supplier: GFS CHEMICALS INC
AMCO Clear® is an EPA approved primary standard for calibration of turbidity sensors.
- Safe, non-toxic and disposable
- easy-to-use with no dilution or resuspension
NIST traceable to particle size/distribution and optical density, and its inherent stability means it does not settle from suspension.
Certifications: Approved by the US EPA for drinking water analysis for turbidity.