Ethanol denatured ≥100% DLI SDA 3A (200 Proof) (denatured with 5% isopropanol and 5% methanol)

Supplier: Decon Labs
Danger

12R1001
71002-398CS 228.86 USD
71002-398
Ethanol denatured ≥100% DLI SDA 3A (200 Proof) (denatured with 5% isopropanol and 5% methanol)
Ethanol denatured

  • Total alcohol: 99.5%
  • Ethyl alcohol: 89-91% (v/v)
  • Denaturant: 4.5-5.5 %(v/v) isopropyl alcohol and 4.0-5.0% (v/v) methyl alcohol


Ordering information: The purchase of Reagent Alcohol in containers larger than 4 liters requires a Letterhead Permit approval by the United States Treasury Tax and Trade Bureau (TTB).

Formula: H₃CCH₂OH
Storage Temperature: Ambient
MDL Number: MFCD00003568
CAS Number: 64-17-5
EINECS: 200-578-6
UN: 1170
ADR: 3,II

Order Now

Specification Test Results

Assay 200 proof
Ethyl alcohol 89-91 (% v/v)
Methyl alcohol 4.0-5.0 (% v/v)
Isopropyl alcohol 4.5-5.5 (% v/v)
Specific gravity @25C 0.782-0.793
Water 0.01 (% v/v) max.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR