Glycerine ≥96% USP

Supplier: Columbus Chemical Industries, Inc

2445RR
71001-988EA 2322 USD
71001-988
Glycerine ≥96% USP
Glycerine
Formula: HOCH₂CH(OH)CH₂OH
Melting Pt: 20 °C (68 °F)
Flash Pt: 160 °C
MDL Number: MFCD00004722
CAS Number: 56-81-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3668cce85af1464db21fd098526252076b567956&vid=86a15453b54e7106c121d32e4e716385003c4507&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance Clear liquid
pH 5.5 - 8
Initial boiling point and boiling range 182 °C at 27 hPa (20 mmHg)
Vapor pressure 0.0033 hPa (0.0025 mmHg) at 50 °C
Vapor density 3.18 (air = 1)
Relative density 1.25 g/mL
Solubility Soluble
Auto-ignition temperature 370 °C (698 °F)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR