Hand Sink, Electronic Eye Faucet, Eagle MHC
Supplier: Eagle Group
HSA-10-FE
71001-944EA 4447.58 USD
71001-944
Hand Sink, Electronic Eye Faucet, Eagle MHC
Furniture Casework
Hand Sink is made up of heavy gauge type 304 stainless steel.
It features splash mounted faucet with electronic eye and basket drain. Electronic eye sensor beam activates water flow and adjusts from 3 to 12". Requires 120-volt, 15-amp electricity. 12-volt adapter.