Ethanol absolute, pure (200 Proof)

Supplier: Decon Labs

2716GEATP
71001-754EA 139.72 USD
71001-754
Ethanol absolute, pure (200 Proof)
Ethanol
Formula: C₂H₅OH
MDL Number: MFCD00003568
CAS Number: 64-17-5
UN: 1170
ADR: 3,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR