SP Bel-Art Tiered Test Tube Rack, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
H18862-0000
71001-744CS 791.49 USD
71001-744
SP Bel-Art Tiered Test Tube Rack, Bel-Art Products, a part of SP
Racks Tube Racks
Heavy-duty polypropylene rack is perfect for organization, storage, and transport of test tubes.
  • Heavy-duty design
  • Polypropylene rack construction
  • Chemically resistant
  • Autoclavable
The 16-place rack holds tubes 10–13 mm in diameter. Heat and chemical resistant, it is autoclavable at 121°C (250°F).