Jumbo Clamp Holder, Ace Glass
Supplier: Ace Glass
11083-51
71001-710EA 101.5 USD
71001-710
Jumbo Clamp Holder, Ace Glass
Clamps General Purpose Clamps
This 316 stainless steel, ninety degree frame clamp holder can be used with all Labjaws extension clamps
It has a 21mm (0.83") grip.