Clamps, Stainless Steel, 3-Prong, Ace Glass
Supplier: Ace Glass
11057-01 11057-03 11057-05
71001-694EA 165.18 USD
71001-694 71001-696 71001-698
Clamps, Stainless Steel, 3-Prong, Ace Glass
Clamps General Purpose Clamps
This multi-purpose, three-prong clamp is manufactured from electro-polished stainless steel for improved corrosion resistance and autoclave sterilization
Features include dual screw adjustments and vinyl sleeved prongs.
Ordering information: Replacement high temperature fiberglass sleeves and stainless steel clamp holders are available for purchase.