Funnel, Powder, Angled, Ace Glass Incorporated
Supplier: Ace Glass
7245-25 7245-15 7245-21 7245-07
71001-676EA 217.83 USD
71001-676 89083-992 89083-994 89083-996
Funnel, Powder, Angled, Ace Glass Incorporated
Funnels Powder Funnels
Powder funnel angled for use in multi-neck flasks
On flasks with angled necks, this design allows the mouth of the funnel to be positioned back at vertical. Fabricated from heavy wall borosilicate glass.