Adapter, Septum Inlet, Ace Glass Incorporated
Supplier: Ace Glass
5110-13 5110-11
71001-504EA 64.41 USD
71001-504 89051-326
Adapter, Septum Inlet, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
This is a sampling adapter with a septum port at the top for handling air-sensitive materials.
  • Septum port at top
  • Bottom [ST] inner joint
  • Septum included
  • Fabricated from borosilicate glass
Featuring a standard taper inner joint at the bottom and septum port at the top. Supplied with a 89064-960 septum.