Purge Adapter, Ace Glass Incorporated
Supplier: Ace Glass
3953-03 3953-05
71001-500EA 147.76 USD
71001-500 71001-502
Purge Adapter, Ace Glass Incorporated
Adapters Adapters, Conical Ground Joints
This glass purge adapter is desgined for use with rotary evaporators.
  • Top spherical socket joint
  • Bottom spherical ball joint
  • Middle tube stopcock or valve
  • Fabricated from borosilicate glass
Top has a spherical 35/25 socket joint for joining to the condenser or trap, and the bottom has a spherical 35/25 ball joint for joining to the receiver flask. Middle tube has either a 4mm PTFE stopcock, or a 2mm valve to allow for purging of the vapor from the receiver flask or for adding purge gas into the flask. Overall length is 75mm.