OSOM® Strep A Test Kit, Sekisui Diagnostics

Supplier: SEKISUI DIAGNOSTIC

141
71001-048CS 6268.14 USD
71001-048
OSOM® Strep A Test Kit, Sekisui Diagnostics
Clinical Diagnostic Supplies Clinical Reagents and Kits

The OSOM® Strep A test kit provides clear, accurate results in as little as five minutes


  • Sensitivity 96%
  • Specificity 98%


The easy-to-read two-color results reduce patient waiting time and allow for quick diagnosis. Specimens are obtained via throat swab. Test kits can be stored for 17 months at room temperature.


Certifications: CLIA-waived. CE marked.


Ordering information: Kit includes two additional tests for external QC testing (50-test kit includes 52 tests).

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR