Tissue Homogenizer, Potter-Elvehjem, Micro, Ace Glass Incorporated
Supplier: Ace Glass
8357-03
71000-520CS 232.31 USD
71000-520
Tissue Homogenizer, Potter-Elvehjem, Micro, Ace Glass Incorporated
Homogenizers Tissue Grinders
This micro size tissue homogenizer is for extremely precise work
Cited in Biochemistry Laboratory, 2nd Edition, by Rodney Boyer, "Tools for the Preparation of Crude Cell Extracts".
This unit is designed for delicate hand operation. A reservoir and pouring lip are incorporated into the design. Fabricated from borosilicate glass.