Flask, Shaker, Three Extra Deep Baffles, Ace Glass Incorporated
Supplier: Ace Glass
3887-17 3887-19 3887-22 3887-11 3887-24 3887-13 3887-05
71000-398CS 450.85 USD
71000-398 71000-410 71000-420 71000-432 71000-440 71000-450 71000-454 71000-466 71000-476
Flask, Shaker, Three Extra Deep Baffles, Ace Glass Incorporated
Flasks Cell Culture Flasks
This shaker flask has a long-style neck with three extra deep baffles to enhance gas transfer when used with a rotary or reciprocating shakers
The long neck reduces splashing, and is designed for polypropylene closures. Fabricated from borosilicate glass.
Ordering information: Closure NOT included.