Corning® 96-Well Clear Round Bottom Polypropylene Microplate, Untreated, Corning
Supplier: Corning
3879
71000-078CS 376.35 USD
71000-078
Corning® 96-Well Clear Round Bottom Polypropylene Microplate, Untreated, Corning
Microplates Cell Culture Plates
PS, clear, with lid.
  • Corning® CellBIND® surface increases wetability for more even and consistent cell attachment
  • Ultra-Low Attachment plates feature a covalently bound hydrogel layer that minimizes cell attachment, protein absorption and cellular activation
  • Sterilized by gamma irradiation
  • Certified non-pyrogenic
Recommended working volume: 75 to 200 µl.