KIMBLE® Westergren Sedimentation Tube, DWK Life Sciences
Supplier: DWK Life Sciences (KIMBLE)
67G70720-035
70720-035CS 912.64 USD
70720-035
KIMBLE® Westergren Sedimentation Tube, DWK Life Sciences
Clinical Diagnostic Supplies Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
ESR tubes measure the rate at which red blood cells settle out of plasma. Westergren sedimentation rate tubes may be used for the standard procedure, by drawing blood in a liquid sodium citrate vacuum tube, or the modified procedure, using a saline-diluted EDTA vacuum tube.
  • Precision-drawn glass and accurate calibrations ensure that the results are accurate with the standard normal values
  • Manufactured from 90 expansion soda lime glass conforming to USP Type III requirements