SP Bel-Art Thistle Tube, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F14751-0000
70690-262CS 668.63 USD
70690-262
SP Bel-Art Thistle Tube, Bel-Art Products, a part of SP
Funnels Filtration Funnels Filter Funnels
Make additions to burettes and narrow neck containers easily.
  • Unbreakable polypropylene plastic construction
  • Chemically resistant
  • Flared top
  • Autoclavable
The flared top thistle tube is made from polypropylene plastic, which is chemical resistant, unbreakable and safe. Steam autoclavable at 121ºC (250ºF). Dimensions are 27 mm (1¹/₁₆") I.D. top with a 41 mm (1⅝") O.D. flare. The 6.4 mm (¹/₄") stem is 254 mm (10") long.