SP Bel-Art Large Powder Funnel, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
H14684-0000
70690-206CS 185.65 USD
70690-206
SP Bel-Art Large Powder Funnel, Bel-Art Products, a part of SP
Funnels Powder Funnels
Ideal for powders and general use, this funnel has an extra wide 1½“ stem exit that allows for extra coarse or clumping powders to easily pass.
  • HDPE construction
  • Exceptional chemical resistance
  • Extra-wide funnel stem exit allows powder clumps to pass
High-density polyethylene provides exceptional chemical resistance to most acids, alkalis and some organic solvents.