SP Bel-Art Gas Dispersion Tube, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F13691-0000
70690-204CS 610.61 USD
70690-204
SP Bel-Art Gas Dispersion Tube, Bel-Art Products, a part of SP
Gas Tubes
High-density polyethylene bell-mouthed fitting with a sealed-in, coarse porosity polyethylene disc at the large end results in smaller bubbles.
  • Sealed-in porosity disc
  • HDPE tube construction
  • Polyethylene disc fabrication
A 30.5 cm long (12") x 6.35 mm ID (¼") low-density polyethylene tube is fitted to the other end and is suitable for insertion into small openings. O.D. of bell is 2.54 cm (1").