SP Bel-Art Thermometer Holder, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F18241-0000
70690-196CS 284.57 USD
70690-196
SP Bel-Art Thermometer Holder, Bel-Art Products, a part of SP
Clamps General Purpose Clamps
Convenient polypropylene holder clips onto the edge of a beaker or other vessel and holds a thermometer vertically near the side of the container for safe and easy reading.
  • Autoclavable
  • Holder fits onto vessels with a wall thickness up to 6.4 mm
Withstands temperatures up to 121 °C (250 °F). The thermometer is held in place by small fingers that will expand to accommodate thermometers ranging from 3.2 mm (1/8") to 11 mm (7/16") in diameter.