SP Bel-Art Stainless Steel Coated Lab Spoon and Spatula, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
H36711-0000
70690-180EA 73.12 USD
70690-180
SP Bel-Art Stainless Steel Coated Lab Spoon and Spatula, Bel-Art Products, a part of SP
Spatulas
This stainless steel sampling spoon and spatula has ends coated with brown, Fluo-Kem® Teflon® FEP that complies with FDA regulations governing direct food contact and provides an anti-stick, corrosion-resistant surface.
  • FEP-coated stainless steel construction
  • Coating provides anti-stick, corrosion-resistant surface
  • Two ends are spatula and spoon
The 4 mm (⁵⁄₃₂") Ø rod is flattened at one end to give an 8×51 mm (⁵⁄₁₆×2") spatula. The opposite end is a 13×25 mm (½×1") spoon.