SP Bel-Art Bulb Pipettor, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
H37888-0000
70690-152CS 5557.74 USD
70690-152
SP Bel-Art Bulb Pipettor, Bel-Art Products, a part of SP
Pipettes Pipette Bulbs
A tapered, silicone rubber seal provides an airtight fit on all size pipettes.
  • Tapered design
  • Silicone rubber seal
  • Autoclavable
An excellent aspirator for use with Westergren or Wintrobe sedimentation tubes. The bulb can be opened, cleaned, and steam autoclaved at 121 °C (250 °F).
Delivery information: Supplied complete with an elastic cord for dedicating the pipettor to a specific reagent bottle.