SP Bel-Art Screw-Clamp Compressor, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F18225-0000
70690-118CS 877.88 USD
70690-118
SP Bel-Art Screw-Clamp Compressor, Bel-Art Products, a part of SP
Clamps Tubing and Sanitary Clamps
Side opening tube clamp permits installation onto tubing without disconnecting from apparatus or interrupting operation.
  • Durable nylon construction
  • Good chemical resistance
  • Easy installation
Constructed of durable nylon for long life and good chemical resistance. Accepts any tubing up to 6.4 mm (1/4") O.D.