SP Bel-Art Slide Holder Strip, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F44542-0000
70690-016EA 22.48 USD
70690-016
SP Bel-Art Slide Holder Strip, Bel-Art Products, a part of SP
Slide Holders
This slide holder features a sloped groove that accepts up to 10 single or double slides standing on end.
  • Polystyrene construction
  • Sloped groove accepts multiple combinations of single and double slides
  • Durable
A front ledge serves as a place to dry slides.