SP Bel-Art Heavy-Duty Pails, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
H16806-0000
70680-542EA 185.13 USD
70680-542 70680-540
SP Bel-Art Heavy-Duty Pails, Bel-Art Products, a part of SP
Buckets
Polyethylene plastic pails have wide rims that hold their shape under heavy loads.
  • Polyethylene construction
  • Plastic-coated metal bail
  • Pronounced spout
  • Graduation markings
  • Recessed hand grip
Features include a heavy-gauge, plastic-coated metal bail and a pronounced spout. Recessed hand grip on the bottom of bucket aids in safe handling and pouring. Graduations are in liters and molded on the interior.