SP Bel-Art Precision Plastic Vernier Calipers, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
H13415-0000
70680-530EA 153.42 USD
70680-530
SP Bel-Art Precision Plastic Vernier Calipers, Bel-Art Products, a part of SP
Calipers
Vernier calipers can be used to measure internal and external dimensions with utmost precision.
  • Superior precise measurements
  • Corrosion resistant
  • Non-conducting
  • Lightweight and easy to handle
Precise measurements can be made with the fine adjustment thumb roller, and reading scales create no parallax
Upper scale is 150 mm with 0.1 mm subdivisions, lower scale is 6 inches with 0.005" subdivisions. Molded-in rigid plastic which won’t scratch like metal, these calipers are non-conducting, corrosion resistant, and lightweight in the hand for easy handling.
Parallel jaws are 40 mm (1½") deep. Inside jaws and round plastic rod depth gauge provide added value.