SP Bel-Art Replica-Plating Device, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F37848-0000
70680-330EA 258.93 USD
70680-330
SP Bel-Art Replica-Plating Device, Bel-Art Products, a part of SP
Microplate and Petri Dish Replicators
Utilizes a locking ring which secures sterile velveteen squares onto a PVC cylinder for easy control of the plating device.
  • PVC cylinder construction
  • Fresh sterile nutrient plate
  • Not autoclavable
  • Easily disinfected
A fresh sterile nutrient plate is then lowered onto the velveteen surface and lifted, leaving a replica of the original colonies in the new dish.
May be disinfected between uses with a brief rinse in 70% ethanol or chlorine bleach. The aluminum ring is 10.2 cm (4") diameter, and the PVC cylinder is 6.9 cm (23/4") high. For use with 90 to 100 mm Petri dishes.
Delivery information: 12 pieces/bag of white velveteen squares is included.