SP Bel-Art Screw Style Tubing Clamp, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F18212-0000
70680-322PK 40.96 USD
70680-322
SP Bel-Art Screw Style Tubing Clamp, Bel-Art Products, a part of SP
Clamps Tubing and Sanitary Clamps
Offers on/off and fine control so accurate that it replaces stopcocks in many applications.
  • Molded acetal plastic construction
  • Very fine, accurate control
  • Autoclavable
  • Resistant to acids, alkalis, and organic solvents
Can be installed without disconnecting tubing or interrupting operation. Molded of Acetal plastic, the clamp is autoclavable and resistant to acids, alkalis and organic solvents. Accepts flexible tubing up to 14 mm (½") O.D.