SP Bel-Art® Tapered Plug Sampler, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
H37826-0000
70680-276EA 244.26 USD
70680-276
SP Bel-Art® Tapered Plug Sampler, Bel-Art Products, a part of SP
Samplers Solid Samplers
A smooth, tapered, stainless sampler for extracting a “plug” of material from bags or blocks of cheese, grease, soap, clay, etc.
  • Stainless steel construction
  • Can easily cut through a bag or box
  • Smooth, tapered design
The sharp end of the satin finish sampler will cut through a bag or box.
Twisting the handle one-half turn will cut out a plug of material 15.2 cm (6”) long tapering from 1.7 cm (¹¹/₁₆”) to 1.3 cm (½”). “T” handle is 7 cm (2¾”).