SP Bel-Art® Liquid Sampler, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
H37827-0000
70680-274EA 395.18 USD
70680-274
SP Bel-Art® Liquid Sampler, Bel-Art Products, a part of SP
Samplers Fluid Samplers
Ideal for viscous liquids, this 50 mL (1.65 oz.) liquid sampler is made for immersion into a can, bottle, tank, vat, etc.
  • Designed for being immersed
  • Compatible with viscous liquids
  • Nickel-plated brass cup construction
  • Finger grips on tubular handle
The nickel-plated brass cup and tubular handle with finger grips are 45 cm (17½”) long
Overall length 50 cm (19¼”), cup diameter 32 mm (1¼”). Cup is not replaceable, and does not detach.
A center rod with finger ring actuates a valve in the bottom of the cup for rapid and convenient emptying when sample is retrieved.