SP Bel-Art Covamat™ Clear Bench Liner, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
H24679-0000
70680-270EA 239.63 USD
70680-270
SP Bel-Art Covamat™ Clear Bench Liner, Bel-Art Products, a part of SP
Surface Protectors Bench Protectors
Clear polyethylene ribbed matting can be used for lining laboratory tables, shelves, trays, etc.
  • Clear polyethylene construction
  • Ribbed design
  • Easily trimmed to fit laboratory surfaces
It is easily trimmed with scissors and also works well under drums or carboys. 0.25 mm (0.010") with 0.7 mm (0.030") thick ribs.