SP Bel-Art Safe-Lab™ Hollow Teflon® PTFE Stoppers for Tapered Joints, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
F20197-1515
70680-172CS 342.17 USD
70680-172 70680-176 70680-180 70680-186
SP Bel-Art Safe-Lab™ Hollow Teflon® PTFE Stoppers for Tapered Joints, Bel-Art Products, a part of SP
Stoppers PE/LDPE/HDPE Stoppers
Hollow Teflon® PTFE stoppers are designed for boiling and distilling flasks with ground joints.
  • PTFE plastic construction
  • Red extraction nut allows safe removal of stoppers
  • Stoppers designed for boiling and distilling flasks
  • Autoclavable
Safe-Lab™ stoppers are easily removed from standard tapered glassware by turning the red extraction nut to apply a safe, gentle, and even force against the mouth of the vessel, helping to eliminate accidents and breakage. Autoclave safe at 121°C (250°F).