Zinc formaldehyde sulfoxylate
Supplier: Spectrum Chemical Mfg. Corp.
|
|
Zinc Formaldehyde Sulfoxylate is used as a discharge agent in textile printing and also as redox catalyst in the manufacturing of polymers.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Storage Temperature:
Ambient |
MDL Number:
MFCD01632686 CAS Number: 24887-06-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...