Zinc formaldehyde sulfoxylate

Supplier: Spectrum Chemical Mfg. Corp.

Z1032-1KG Z1032-250GM Z1032-5KG
700016-268EA 1017.55 USD
700016-268 700016-270 700016-272
Zinc formaldehyde sulfoxylate
Zinc formaldehyde sulfoxylate

Zinc Formaldehyde Sulfoxylate is used as a discharge agent in textile printing and also as redox catalyst in the manufacturing of polymers.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Storage Temperature: Ambient
MDL Number: MFCD01632686
CAS Number: 24887-06-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR