Wheat germ oil

Supplier: Spectrum Chemical Mfg. Corp.

WH110-500MLG WH110-4LT
700016-222EA 175.45 USD
700016-222 700016-220
Wheat germ oil
Wheat germ oil

Wheat Germ Oil is an emollient and used in cosmetics because of its high vitamin e content.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Storage Temperature: Ambient
MDL Number: MFCD00132584
CAS Number: 8006-95-9

Order Now

Specification Test Results

Specific Gravity @ 20°C 0.900 - 0.940
Saponification Value 180 - 195
Iodine Value 120 - 140
Moisture max. 0.2 %
Free Fatty Acids max. 1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR