Triglycerine

Supplier: Spectrum Chemical Mfg. Corp.

T1344-250ML
700015-960EA 200.2 USD
700015-960
Triglycerine
Triglycerine

Triglycerine is used as a skin conditioning agent and humectant.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₉H₂₀O₇
Density: 1.30
Storage Temperature: Ambient
MDL Number: MFCD00084755
CAS Number: 56090-54-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR