Sodium hexametaphosphate

Supplier: Spectrum Chemical Mfg. Corp.

S2603-1KG
700015-840EA 639.88 USD
700015-840
Sodium hexametaphosphate
Sodium hexametaphosphate

Sodium Hexametaphosphate is used as a sequestrant and a food additive, as well as a dispersing agent to break down soil types. It is typically a mixture of polymeric metaphosphates. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Formula: (NaO₃P)n
Melting Pt: 610 °C
Density: 1.25 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134118
CAS Number: 68915-31-1
EINECS: 233-343-1

Order Now

Specification Test Results

Assay 66.5% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR