Sodium hexadecyl sulfate

Supplier: Spectrum Chemical Mfg. Corp.

S1422-100GM S1422-25GM S1422-500GM
700015-734EA 300.5 USD
700015-734 700015-736 700015-738
Sodium hexadecyl sulfate
Sodium hexadecyl sulphate

Sodium n-Hexadecyl Sulfate is used as an anionic cleansing and emulsifying surfactant.


Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formula: C₁₆H₃₃NaO₄S
Storage Temperature: Ambient
MDL Number: MFCD00047766
CAS Number: 1120-01-0
EINECS: 214-292-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR