Cationic starch N 0.3 %

Supplier: Spectrum Chemical Mfg. Corp.

S1189-100MG S1189-250MG S1189-1GM
700015-656EA 183.15 USD
700015-656 700015-660 700015-658
Cationic starch N 0.3 %
Cationic starch

Cationic Starch, Nitrogen 0.3 Percent is a carbohydrate and polysaccharide that has a large number of glucose units that are linked by glycosidic bonds that has been modified by reaction with a quaternary ammonium salt. These products are used to increase the dry strength and emulsification of sizing agents.

Storage Temperature: Ambient
CAS Number: 56780-58-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR