Rose water artificial
Supplier: Spectrum Chemical Mfg. Corp.Specification Test Results
Specific Gravity @ 25°C | 0.990 - 1.010 |
Solubility in Water | To Pass Test |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...