Potassium tripolyphosphate
Supplier: SPECTRUM CHEMICAL MFG CORP
Synonyms:
Potassium triphosphate
P1669-2.5KG
P1669-12KG
P1669-500GM
700015-140EA
329.65
USD
700015-140
700015-138
700015-142
Potassium tripolyphosphate
Potassium tripolyphosphate
Potassium Tripolyphosphate is used a texturizing agent and acidity regulator.
Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.
Formula:
K₅O₁₀P₃ Storage Temperature: Ambient |
MDL Number:
MFCD00054129 CAS Number: 13845-36-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...